Ethoxyacetic acid ≥98.0%
Supplier: TCI America
Synonyms:
2-Ethoxyacetic acid
E0377-25G
E0377-500G
TCE0377-25GEA
53
USD
TCE0377-25G
TCE0377-500G
Ethoxyacetic acid ≥98.0%
Ethoxyacetic acid
CAS Number: 627-03-2
MDL Number: MFCD00004310
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 97
Specific Gravity (20/20): 1.10
Formula:
C₄H₈O₃ Boiling Pt: 207 °C Density: 1.10 (20 °C) Flash Pt: 97 °C |
MDL Number:
MFCD00004310 CAS Number: 627-03-2 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...