2-Ethyl-3-methylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0361-25ML E0361-5ML
TCE0361-25MLEA 65 USD
TCE0361-25ML TCE0361-5ML
2-Ethyl-3-methylpyrazine ≥98.0% (by GC, titration analysis)
2-Ethyl-3-methylpyrazine

CAS Number: 15707-23-0
MDL Number: MFCD00006150
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 57
Specific Gravity (20/20): 0.99

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 70 °C
Density: 0.99 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00006150
CAS Number: 15707-23-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a7066c404c27bf772883dbb3fea4fcf57b5b6a&vid=86a7066c404c27bf772883dbb3fea4fcf57b5b6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR