Ethyl methyl sulfide ≥98.0% (by GC)

Supplier: TCI America

E0143-100ML E0143-25ML
TCE0143-100MLEA 107 USD
TCE0143-100ML TCE0143-25ML
Ethyl methyl sulfide ≥98.0% (by GC)
Ethyl methyl sulphide

CAS Number: 624-89-5
MDL Number: MFCD00009268
Molecular Formula: C3H8S
Molecular Weight: 76.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 66
Flash Point (°C): -15
Specific Gravity (20/20): 0.84

Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 66 °C
Density: 0.84 (20 °C)
Flash Pt: -15 °C
MDL Number: MFCD00009268
CAS Number: 624-89-5
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45be1bc827a1233cfbff9a70179aa3b26dda38ce&vid=45be1bc827a1233cfbff9a70179aa3b26dda38ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR