Ergosterol ≥95.0% (by GC)

Supplier: TCI America

E0018-25G E0018-5G
TCE0018-25GEA 207.52 USD
TCE0018-25G TCE0018-5G
Ergosterol ≥95.0% (by GC)
Ergosterol

CAS Number: 57-87-4
MDL Number: MFCD00003623
Molecular Formula: C28H44O
Molecular Weight: 396.66
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 250
Melting point (°C): 159
Specific rotation [a]20/D: -113 deg (C=1, THF)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Anti-osteoporosis Ingredients, Vitamins, Phytosteroids, Steroids
Other Product Names / Synonyms: Provitamin D2

Formula: C₂₈H₄₄O
Boiling Pt: 250 °C
Melting Pt: 159 °C
MDL Number: MFCD00003623
CAS Number: 57-87-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fb6224aaf8fc21dfeda6c6d7f987fdca0f96ffd&vid=4113f5a860c6623483b49ed94552269f668a467b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR