3,3-Dimethylbutyraldehyde ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 3,3-Dimethylbutanal

D4098-25ML D4098-5ML
TCD4098-25MLEA 245 USD
TCD4098-25ML TCD4098-5ML
3,3-Dimethylbutyraldehyde ≥97.0% (by GC)
3,3-Dimethylbutyraldehyde

CAS Number: 2987-16-8
MDL Number: MFCD00042807
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 13
Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: 3,3-Dimethylbutanal

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 104 °C
Density: 0.80 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00042807
CAS Number: 2987-16-8
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6191a77bed9135da8dfb3d9f416f0eaa66feac1&vid=a6191a77bed9135da8dfb3d9f416f0eaa66feac1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR