2,2-Difluoro-1,3-benzodioxole ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,2-Difluorobenzo[d][1,3]dioxole
D4051-25G
D4051-5G
TCD4051-25GEA
118.2
USD
TCD4051-25G
TCD4051-5G
2,2-Difluoro-1,3-benzodioxole ≥98.0% (by GC)
2,2-Difluoro-1,3-benzodioxole
CAS Number: 1583-59-1
MDL Number: MFCD00236217
Molecular Formula: C7H4F2O2
Molecular Weight: 158.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 32
Specific Gravity (20/20): 1.32
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,2-(Difluoromethylenedioxy)benzene
Formula:
C₇H₄F₂O₂ MW: 158.1 g/mol Density: 1.32 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00236217 CAS Number: 1583-59-1 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...