2,4-Dichlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,4-Dichlorobenzene-1-sulphonyl chloride

D4037-25G D4037-5G
TCD4037-25GEA 135 USD
TCD4037-25G TCD4037-5G
2,4-Dichlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
2,4-Dichlorobenzenesulphonyl chloride

CAS Number: 16271-33-3
MDL Number: MFCD00052712
Molecular Formula: C6H3Cl3O2S
Molecular Weight: 245.50
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 57

Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Melting Pt: 57 °C
MDL Number: MFCD00052712
CAS Number: 16271-33-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bb97ed3f8390619b555431a8c6492b50a45c26a&vid=7bb97ed3f8390619b555431a8c6492b50a45c26a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR