2,5-Difluorobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (2,5-Difluorophenyl)methanamine

D4034-5G D4034-1G
TCD4034-5GEA 281 USD
TCD4034-5G TCD4034-1G
2,5-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
2,5-Difluorobenzylamine

CAS Number: 85118-06-5
MDL Number: MFCD00010143
Molecular Formula: C7H7F2N
Molecular Weight: 143.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 76
Specific Gravity (20/20): 1.23

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 78 °C
Density: 1.23 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00010143
CAS Number: 85118-06-5
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50faa5eb572137c10d371672c18413da2ce7a51&vid=c50faa5eb572137c10d371672c18413da2ce7a51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR