3,5-Dimethoxybenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,5-Dimethoxybenzyl amine

D4025-25G D4025-5G
TCD4025-25GEA 335 USD
TCD4025-25G TCD4025-5G
3,5-Dimethoxybenzylamine ≥98.0% (by GC, titration analysis)
3,5-Dimethoxybenzylamine

CAS Number: 34967-24-3
MDL Number: MFCD00052813
Molecular Formula: C9H13NO2
Molecular Weight: 167.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 39
Flash Point (°C): 113
Lambda max.: 281 nm (EtOH)Storage Temperature: 0-10°C

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 96 °C
Melting Pt: 39 °C
Flash Pt: 113 °C
MDL Number: MFCD00052813
CAS Number: 34967-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54511551efd75c11eea2910d7b46fc1f03a4bbe9&vid=54511551efd75c11eea2910d7b46fc1f03a4bbe9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR