3,4-Dimethoxybenzylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Veratrylamine

D4024-25G D4024-5G
TCD4024-25GEA 228 USD
TCD4024-25G TCD4024-5G
3,4-Dimethoxybenzylamine ≥97.0% (by GC, titration analysis)
3,4-Dimethoxybenzylamine

CAS Number: 5763-61-1
MDL Number: MFCD00008116
Molecular Formula: C9H13NO2
Molecular Weight: 167.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 113
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Veratrylamine

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 156 °C
Density: 1.12 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008116
CAS Number: 5763-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03792fbac004dca0ad133eefb8111ea0a8df0481&vid=03792fbac004dca0ad133eefb8111ea0a8df0481&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR