Diethyl-2,5-dibromoadipate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Diethyl 2,5-dibromohexanedioate, 2,5-Dibromoadipic acid diethyl ester

D3965-25G D3965-5G
TCD3965-25GEA 112 USD
TCD3965-25G TCD3965-5G
Diethyl-2,5-dibromoadipate ≥98.0% (by GC)
Diethyl-2,5-dibromoadipate

CAS Number: 869-10-3
MDL Number: MFCD00075379
Molecular Formula: C10H16Br2O4
Molecular Weight: 360.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 134
Melting point (°C): 66


Other Product Names / Synonyms: 2,5-Dibromoadipic Acid Diethyl Ester

Formula: C₁₀H₁₆Br₂O₄
MW: 360.04 g/mol
Boiling Pt: 134 °C
Melting Pt: 66 °C
MDL Number: MFCD00075379
CAS Number: 869-10-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63303a8e597951722d12230d0389e1541726c6ac&vid=14d8fbc0d5340a9e80633c74812145bd5b8dd1fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR