1,4-Dibromo-2,5-bis(bromomethyl)benzene ≥97.0%

Supplier: TCI America

D3961-1G D3961-5G
TCD3961-1GEA 93 USD
TCD3961-1G TCD3961-5G
1,4-Dibromo-2,5-bis(bromomethyl)benzene ≥97.0%
1,4-Dibromo-2,5-bis(bromomethyl)benzene

CAS Number: 35335-16-1
MDL Number: MFCD03411745
Molecular Formula: C8H6Br4
Molecular Weight: 421.75
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: alpha,alpha,2,5-Tetrabromo-p-xylene

Formula: C₈H₆Br₄
MW: 421.7500 g/mol
Melting Pt: 160 °C
MDL Number: MFCD03411745
CAS Number: 35335-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9c99ef4d043d51d7f4674f932befabdfc104b19&vid=d9c99ef4d043d51d7f4674f932befabdfc104b19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR