Decylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: n-Decylphosphonic acid, 1-Decylphosphonic acid

D3884-5G D3884-1G
TCD3884-5GEA 234 USD
TCD3884-5G TCD3884-1G
Decylphosphonic acid ≥98.0% (by titrimetric analysis)
Decylphosphonic acid

CAS Number: 6874-60-8
MDL Number: MFCD00015830
Molecular Formula: C10H23O3P
Molecular Weight: 222.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 102

Formula: C₁₀H₂₃O₃P
MW: 222.26 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00015830
CAS Number: 6874-60-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce05c52dd7ecf34858d167d1e08725521c0fd50&vid=8ce05c52dd7ecf34858d167d1e08725521c0fd50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR