Dibromoisocyanuric acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: DBI, 1,3-Dibromo-1,3,5-triazine-2,4,6-trione

D3753-25G D3753-5G
TCD3753-25GEA 254.2 USD
TCD3753-25G TCD3753-5G
Dibromoisocyanuric acid ≥97.0% (by titrimetric analysis)
Dibromoisocyanuric acid

CAS Number: 15114-43-9
MDL Number: MFCD00463941
Molecular Formula: C3HBr2N3O3
Molecular Weight: 286.87
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 309
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: 1,3-Dibromo-1,3,5-triazine-2,4,6-trione, DBI

Formula: C₃HBr₂N₃O₃
MW: 286.87 g/mol
Melting Pt: 309 °C
MDL Number: MFCD00463941
CAS Number: 15114-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c000441d5ae63f2bc5ab60e61ccb58308ea1f674&vid=c000441d5ae63f2bc5ab60e61ccb58308ea1f674&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR