2,5-Difluorophenyl isocyanate ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 39718-32-6
MDL Number: MFCD00013843
Molecular Formula: C7H3F2NO
Molecular Weight: 155.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 54
Flash Point (°C): 56
Specific Gravity (20/20): 1.33
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isocyanic Acid 2,5-Difluorophenyl Ester
Formula:
C₇H₃F₂NO Boiling Pt: 54 °C Density: 1.33 (20 °C) Flash Pt: 56 °C |
MDL Number:
MFCD00013843 CAS Number: 39718-32-6 UN: 3080 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...