2,5-Dimethoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America

D3659-1G D3659-5G
TCD3659-1GEA 26 USD
TCD3659-1G TCD3659-5G
2,5-Dimethoxybenzonitrile ≥98.0% (by GC)
2,5-Dimethoxybenzonitrile

CAS Number: 5312-97-0
MDL Number: MFCD00016375
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 83

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00016375
CAS Number: 5312-97-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c51876edf5a733bdd2cc4d2cd68e12257124a899&vid=623e814f2bbb678cc3a44d80e361d099277c1d52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR