1,3-Diiodo-5,5-dimethylhydantoin ≥97.0%
Supplier: TCI America
Synonyms:
DIH
D3657-25G
D3657-5G
TCD3657-25GEA
330
USD
TCD3657-25G
TCD3657-5G
1,3-Diiodo-5,5-dimethylhydantoin ≥97.0%
1,3-Diiodo-5,5-dimethylhydantoin
CAS Number: 2232-12-4
MDL Number: MFCD00020867
Molecular Formula: C5H6I2N2O2
Molecular Weight: 379.92
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 198
Flash Point (°C): 115
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: DIH
Formula:
C₅H₆I₂N₂O₂ MW: 379.92 g/mol Melting Pt: 198 °C Flash Pt: 115 °C |
MDL Number:
MFCD00020867 CAS Number: 2232-12-4 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...