Diisopropyl(bromomethyl)boronate ≥95.0%
Supplier: TCI America
Synonyms:
(Bromomethyl)boronic Acid Diisopropyl Ester
D3649-1G
D3649-5G
TCD3649-1GEA
138
USD
TCD3649-1G
TCD3649-5G
Diisopropyl(bromomethyl)boronate ≥95.0%
Diisopropyl(bromomethyl)boronate
CAS Number: 137297-49-5
MDL Number: MFCD01631222
Molecular Formula: C7H16BBrO2
Molecular Weight: 222.92
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: (Bromomethyl)boronic Acid Diisopropyl Ester
Formula:
C₇H₁₆BBrO₂ MW: 222.92 g/mol |
MDL Number:
MFCD01631222 CAS Number: 137297-49-5 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...