Chloro(di-tert-butyl)phosphine ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
Di-tert-butylchlorophosphine, Di-t-butylchlorophosphine
D3632-25G
D3632-5G
TCD3632-25GEA
256.64
USD
TCD3632-25G
TCD3632-5G
Chloro(di-tert-butyl)phosphine ≥95.0% (by GC)
Chloro(di-tert-butyl)phosphine
CAS Number: 13716-10-4
MDL Number: MFCD00008815
Molecular Formula: C8H18ClP
Molecular Weight: 180.66
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 72
Melting point (°C): 3
Flash Point (°C): 62
Specific Gravity (20/20): 0.95
Formula:
C₈H₁₈ClP MW: 180.66 g/mol Boiling Pt: 72 °C Melting Pt: 3 °C Density: 0.95 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00008815 CAS Number: 13716-10-4 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...