2,6-Dichloro-4-fluoroaniline ≥97.0% (by GC, total nitrogen)

Supplier: TCI America

D3625-1G D3625-5G
TCD3625-1GEA 33.5 USD
TCD3625-1G TCD3625-5G
2,6-Dichloro-4-fluoroaniline ≥97.0% (by GC, total nitrogen)
2,6-Dichloro-4-fluoroaniline

CAS Number: 344-19-4
MDL Number: MFCD00142845
Molecular Formula: C6H4Cl2FN
Molecular Weight: 180.00
Purity/Analysis Method: >97.0% (GC,N)
Form: Crystal
Melting point (°C): 56

Formula: C₆H₄Cl₂FN
Melting Pt: 56 °C
MDL Number: MFCD00142845
CAS Number: 344-19-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1aca3d0c47b71595a1359de7e14020790d0acbe&vid=a1aca3d0c47b71595a1359de7e14020790d0acbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR