10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2,2'-Iminodibenzyl-5-carbonyl Chloride
D3565-25G
D3565-5G
TCD3565-25GEA
124
USD
TCD3565-25G
TCD3565-5G
10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl chloride ≥98.0% (by HPLC, titration analysis)
10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl chloride
CAS Number: 33948-19-5
MDL Number: MFCD00012314
Molecular Formula: C15H12ClNO
Molecular Weight: 257.72
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 122
Other Product Names / Synonyms: 2,2'-Iminodibenzyl-5-carbonyl Chloride
Formula:
C₁₅H₁₂ClNO MW: 257.72 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD00012314 CAS Number: 33948-19-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...