2,3-Dihydro-1-benzofuran-5-sulfonoylchloride ≥98.0% (by GC)
Supplier: TCI America
D3553-1G
D3553-5G
TCD3553-1GEA
85
USD
TCD3553-1G
TCD3553-5G
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride ≥98.0% (by GC)
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride
CAS Number: 115010-11-2
MDL Number: MFCD02677703
Molecular Formula: C8H7ClO3S
Molecular Weight: 218.65
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85
Storage Temperature: 0-10°C
Formula:
C₈H₇ClO₃S MW: 218.66 g/mol Melting Pt: 85 °C |
MDL Number:
MFCD02677703 CAS Number: 115010-11-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...