3,5-Dimethylbenzoyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D3550-25G D3550-5G
TCD3550-25GEA 43.76 USD
TCD3550-25G TCD3550-5G
3,5-Dimethylbenzoyl chloride ≥97.0% (by GC, titration analysis)
3,5-Dimethylbenzoyl chloride

CAS Number: 6613-44-1
MDL Number: MFCD02313742
Molecular Formula: C9H9ClO
Molecular Weight: 168.62
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 127
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: m-Xylene-5-carbonyl Chloride

Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 127 °C
Density: 1.14 (20 °C)
MDL Number: MFCD02313742
CAS Number: 6613-44-1
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49dde89c47b6aad3edddbc8b7d545b1e92c77c71&vid=49dde89c47b6aad3edddbc8b7d545b1e92c77c71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR