2,4-Dichloro-3-ethyl-6-nitrophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3510-25G D3510-5G
TCD3510-25GEA 267.8 USD
TCD3510-25G TCD3510-5G
2,4-Dichloro-3-ethyl-6-nitrophenol ≥98.0% (by GC, titration analysis)
2,4-Dichloro-3-ethyl-6-nitrophenol

CAS Number: 99817-36-4
MDL Number: MFCD00270764
Molecular Formula: C8H7Cl2NO3
Molecular Weight: 236.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 48

Formula: C₈H₇Cl₂NO₃
MW: 236.05 g/mol
Melting Pt: 48 °C
MDL Number: MFCD00270764
CAS Number: 99817-36-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f7ea4c998a8bfb1f29ce24a51baf5f2768994a9&vid=7f7ea4c998a8bfb1f29ce24a51baf5f2768994a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR