2-Chloromethyl-m-xylene ≥98.0%
Supplier: TCI America
Synonyms:
α-Chloro-1,2,6-trimethylbenzene, 1-(Chloromethyl)-2,6-dimethylbenzene, 2,6-Dimethylbenzyl chloride, 2-(Chloromethyl)-1,3-dimethylbenzene
D3501-25G
D3501-5G
TCD3501-25GEA
262.6
USD
TCD3501-25G
TCD3501-5G
2-Chloromethyl-m-xylene ≥98.0%
2-Chloromethyl-m-xylene
CAS Number: 5402-60-8
MDL Number: MFCD01310815
Molecular Formula: C9H11Cl
Molecular Weight: 154.64
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 100
Freezing point (°C): 33
Other Product Names / Synonyms: 1-Chloromethyl-2,6-dimethylbenzene
Formula:
C₉H₁₁Cl Boiling Pt: 100 °C |
MDL Number:
MFCD01310815 CAS Number: 5402-60-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...