2,3-Difluorobenzonitrile ≥97.0% (by GC)

Supplier: TCI America

D3500-1G D3500-5G
TCD3500-1GEA 17 USD
TCD3500-1G TCD3500-5G
2,3-Difluorobenzonitrile ≥97.0% (by GC)
2,3-Difluorobenzonitrile

CAS Number: 21524-39-0
MDL Number: MFCD00009976
Molecular Formula: C7H3F2N
Molecular Weight: 139.10
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Flash Point (°C): 75
Specific Gravity (20/20): 1.26

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 188 °C
Density: 1.26 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00009976
CAS Number: 21524-39-0
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61a34dba8714cc7f3cf8cb94a1b769cacfc50ef1&vid=84439041cd2cdf540c60ba24451dda834dd1edfa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR