3,3-Dimethylcyclohexanone ≥97.0% (by GC)

Supplier: TCI America

D3495-1G D3495-5G
TCD3495-1GEA 28 USD
TCD3495-1G TCD3495-5G
3,3-Dimethylcyclohexanone ≥97.0% (by GC)
3,3-Dimethylcyclohexanone

CAS Number: 2979-19-3
MDL Number: MFCD00040178
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 59
Specific Gravity (20/20): 0.91

Formula: C₈H₁₄O
Boiling Pt: 175 °C
Density: 0.91 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00040178
CAS Number: 2979-19-3
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7aadc36db9b62ba90be1c48cf311f7e3a9960ea2&vid=7aadc36db9b62ba90be1c48cf311f7e3a9960ea2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR