6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)-diylbis(methylammonium)tetrafluoroborate [=(R,R)-TaDiAS-2nd] ≥97.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: (R,R)-TaDiAS-2nd

D3475-1G D3475-200MG
TCD3475-1GEA 659 USD
TCD3475-1G TCD3475-200MG
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)-diylbis(methylammonium)tetrafluoroborate [=(R,R)-TaDiAS-2nd] ≥97.0% (by total nitrogen basis)
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)-diylbis(methylammonium)tetrafluoroborate [=(R,R)-TaDiAS-2nd]

Molecular Weight: 1000.82
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Molecular Formula: C58H70B2F8N2O2


Other Product Names / Synonyms: (R,R)-TaDiAS-2nd

Formula: C₅₈H₇₀B₂F₈N₂O₂
MW: 1000.82 g/mol
CAS Number: 2010983-27-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dd21d453d02cc058b1aa4ca3482273b6622d6ee&vid=2dd21d453d02cc058b1aa4ca3482273b6622d6ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR