3,5-Dimethylphenyl isocyanate ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 54132-75-1
MDL Number: MFCD00013868
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 84
Specific Gravity (20/20): 1.04
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Isocyanic Acid 3,5-Dimethylphenyl Ester
Formula:
C₉H₉NO Boiling Pt: 205 °C Density: 1.04 (20 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00013868 CAS Number: 54132-75-1 UN: 2206 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...