3,5-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3407-25G D3407-5G
TCD3407-25GEA 138.1 USD
TCD3407-25G TCD3407-5G
3,5-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
3,5-Difluorobenzoyl chloride

CAS Number: 129714-97-2
MDL Number: MFCD00010309
Molecular Formula: C7H3ClF2O
Molecular Weight: 176.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 69
Specific Gravity (20/20): 1.41

Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Density: 1.41 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00010309
CAS Number: 129714-97-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82063875b0f67cadec2b4f8ac5a06206ad88fccd&vid=82063875b0f67cadec2b4f8ac5a06206ad88fccd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR