Dimethoxymethylvinylsilane ≥94.0% (by GC)

Supplier: TCI America

Synonyms: Dimethoxy(methyl)(vinyl)silane

D3386-250G D3386-25G
TCD3386-250GEA 179.9 USD
TCD3386-250G TCD3386-25G
Dimethoxymethylvinylsilane ≥94.0% (by GC)
Dimethoxymethylvinylsilane

CAS Number: 16753-62-1
MDL Number: MFCD00129170
Molecular Formula: C5H12O2Si
Molecular Weight: 132.23
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 3
Specific Gravity (20/20): 0.89

Formula: C₅H₁₂O₂Si
MW: 132.23 g/mol
Boiling Pt: 104 °C
Density: 0.89 (20 °C)
Flash Pt: 3 °C
MDL Number: MFCD00129170
CAS Number: 16753-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d82b29e6e4456dc262472806a20e50dee5f1b4d&vid=6d82b29e6e4456dc262472806a20e50dee5f1b4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR