2,6-Dibromophenol 98,0

Supplier: TCI America

Synonyms: 1,3-Dibromo-2-hydroxybenzene

D3384-25G D3384-5G
TCD3384-25GEA 163.2 USD
TCD3384-25G TCD3384-5G
2,6-Dibromophenol 98,0
2,6-Dibromophenol

CAS Number: 608-33-3
MDL Number: MFCD00002152
Molecular Formula: C6H4Br2O
Molecular Weight: 251.91
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 56
Lambda max.: 286 nm (EtOH)

Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Boiling Pt: 255
Melting Pt: 56 °C
MDL Number: MFCD00002152
CAS Number: 608-33-3
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f60cad59d4b04a48666e3fe30008051a569d0b1b&vid=aa45193e35adb93d382931fd68740baa572b54d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR