Diethylbenzene (mixture of isomers) ≥98.0%

Supplier: TCI America

D3378-25ML D3378-500ML
TCD3378-25MLEA 23 USD
TCD3378-25ML TCD3378-500ML
Diethylbenzene (mixture of isomers) ≥98.0%
Diethylbenzene (mixture of isomers)

CAS Number: 25340-17-4
MDL Number: MFCD00792900
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Melting point (°C): -31
Flash Point (°C): 55
Specific Gravity (20/20): 0.87

Formula: C₁₀H₁₄
Boiling Pt: 184 °C
Melting Pt: –31 °C
Density: 0.87 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00792900
CAS Number: 25340-17-4
UN: 2049
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR