3,4-Difluorobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3373-1G D3373-5G
TCD3373-1GEA 49 USD
TCD3373-1G TCD3373-5G
3,4-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
3,4-Difluorobenzylamine

CAS Number: 72235-53-1
MDL Number: MFCD00010145
Molecular Formula: C7H7F2N
Molecular Weight: 143.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 79
Specific Gravity (20/20): 1.22

Formula: C₇H₇F₂N
Density: 1.22 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00010145
CAS Number: 72235-53-1
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ddf88e02b1fb633020bf1f8c5fc601ddbb081a6&vid=6ddf88e02b1fb633020bf1f8c5fc601ddbb081a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR