cis-Platin

Supplier: TCI America

Synonyms: Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride

D3371-100MG D3371-1G
TCD3371-100MGEA 83.19 USD
TCD3371-100MG TCD3371-1G
cis-Platin
cis-Platin

CAS Number: 15663-27-1
Molecular Formula: H6Cl2N2Pt
Molecular Weight: 300.05
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 270


Other Product Names / Synonyms: Cisplatin, cis-Diamminedichloroplatinum(II), cis-Platinum(II) Diammine Dichloride

Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 270 °C
MDL Number: MFCD00011623
CAS Number: 15663-27-1
UN: 3288
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=302db6fac7fb4af592838f6375af8fe64dcba2c6&vid=302db6fac7fb4af592838f6375af8fe64dcba2c6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR