2,6-Difluoroanisole ≥97.0% (by GC)
Supplier: TCI America
D3360-1G
D3360-5G
TCD3360-1GEA
20
USD
TCD3360-1G
TCD3360-5G
2,6-Difluoroanisole ≥97.0% (by GC)
2,6-Difluoroanisole
CAS Number: 437-82-1
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 50
Specific Gravity (20/20): 1.21
Formula:
C₇H₆F₂O Boiling Pt: 50 °C Density: 1.21 (20 °C) |
MDL Number:
MFCD00142846 CAS Number: 437-82-1 UN: 1993 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...