2,6-Dichloro-3-cyano-4-picoline ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,6-Dichloro-4-methylnicotinonitrile, 2,6-dichloro-4-methylpyridine-3-carbonitrile, 2,6-Dichloro-3-cyano-4-methylpyridine

D3346-25G D3346-5G
TCD3346-25GEA 57 USD
TCD3346-25G TCD3346-5G
2,6-Dichloro-3-cyano-4-picoline ≥98.0% (by GC)
2,6-Dichloro-3-cyano-4-picoline

CAS Number: 875-35-4
MDL Number: MFCD00052633
Molecular Formula: C7H4Cl2N2
Molecular Weight: 187.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 111


Other Product Names / Synonyms: 2,6-Dichloro-4-methylnicotinonitrile

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
Boiling Pt: 118 °C
Melting Pt: 111 °C
MDL Number: MFCD00052633
CAS Number: 875-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=830e36b25e79324e44445e95c58da12c588acdcf&vid=830e36b25e79324e44445e95c58da12c588acdcf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR