Diethyl (4-cyanobenzyl)phosphonate ≥98.0%

Supplier: TCI America

Synonyms: Diethyl 4-cyanobenzylphosphonate

D3323-25G D3323-5G
TCD3323-25GEA 446 USD
TCD3323-25G TCD3323-5G
Diethyl (4-cyanobenzyl)phosphonate ≥98.0%
Diethyl (4-cyanobenzyl)phosphonate

CAS Number: 1552-41-6
Molecular Formula: C12H16NO3P
Molecular Weight: 253.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 169
Melting point (°C): 31


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (4-Cyanobenzyl)phosphonic Acid Diethyl Ester, 4-(Diethylphosphonomethyl)benzonitrile

Formula: C₁₂H₁₆NO₃P
Boiling Pt: 169 °C
Melting Pt: 31 °C
MDL Number: MFCD08274488
CAS Number: 1552-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5c1954a392a2e476ce9c4a23e7651e8c95d3f29&vid=b5c1954a392a2e476ce9c4a23e7651e8c95d3f29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR