3,5-Dichlorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
D3261-25G
D3261-5G
TCD3261-25GEA
118.2
USD
TCD3261-25G
TCD3261-5G
3,5-Dichlorobenzonitrile ≥98.0% (by GC)
3,5-Dichlorobenzonitrile
CAS Number: 6575-00-4
Molecular Formula: C7H3Cl2N
Molecular Weight: 172.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68
Formula:
C₇H₃Cl₂N Melting Pt: 68 °C |
MDL Number:
MFCD00001800 CAS Number: 6575-00-4 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...