Dichloro(diethylamino)phosphine ≥98.0% (by GC)

Supplier: TCI America

D3244-25G D3244-5G
TCD3244-25GEA 331.6 USD
TCD3244-25G TCD3244-5G
Dichloro(diethylamino)phosphine ≥98.0% (by GC)
Dichloro(diethylamino)phosphine

CAS Number: 1069-08-5
Molecular Formula: C4H10Cl2NP
Molecular Weight: 174.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 83
Specific Gravity (20/20): 1.17
Storage Temperature: 0-10°C


Other Product Names / Synonyms: (Diethylamino)phosphonous Dichloride

Formula: C₄H₁₀Cl₂NP
Boiling Pt: 189 °C
Density: 1.17 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00013614
CAS Number: 1069-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b77834a04e8fb7f1e71d2f7bfa9485077ecb0d0&vid=4b77834a04e8fb7f1e71d2f7bfa9485077ecb0d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR