N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine ≥99.0% (by HPLC, total nitrogen), purified by sublimation

Supplier: TCI America

Synonyms: 4,4'-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, N,N'-Diphenyl-N,N'-di(m-tolyl)benzidine, TPD, N,N'-Bis(3-methylphenyl)-N,N'-diphenylbenzidine

D3236-1G D3236-5G
TCD3236-1GEA 82 USD
TCD3236-1G TCD3236-5G
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine ≥99.0% (by HPLC, total nitrogen), purified by sublimation
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine

CAS Number: 65181-78-4
MDL Number: MFCD00144965
Molecular Formula: C38H32N2
Molecular Weight: 516.69
Purity/Analysis Method: >99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 170


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Highly Purified Reagents
Other Product Names / Synonyms: N,N'-Bis(3-methylphenyl)-N,N'-diphenylbenzidine, 4,4'-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, TPD

Formula: C₃₈H₃₂N₂
MW: 516.6741 g/mol
Melting Pt: 170 °C
MDL Number: MFCD00144965
CAS Number: 65181-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b1954f6bc16f6cb29a4a2ddd1145ea85cbe5c0a&vid=7f052c40b22b50c3e2e2aa8aeb7848091b39ad00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR