Di-tert-butylsilyl-bis(trifluoromethanesulfonate) ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Di-tert-butylsilyl Ditriflate, Di-tert-butylbis(trifluoromethanesulphonyloxy)silane

D3135-1G D3135-5G
TCD3135-1GEA 45.76 USD
TCD3135-1G TCD3135-5G
Di-tert-butylsilyl-bis(trifluoromethanesulfonate) ≥97.0% (by titrimetric analysis)
Di-tert-butylsilyl-bis(trifluoromethanesulfonate)

CAS Number: 85272-31-7
Molecular Formula: C10H18F6O6S2Si
Molecular Weight: 440.44
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Flash Point (°C): 90
Specific Gravity (20/20): 1.36


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Di-tert-butylbis(trifluoromethanesulfonyloxy)silane, Di-tert-butylsilyl Ditriflate

Formula: C₁₀H₁₈F₆O₆S₂Si
MW: 440.44 g/mol
Density: 1.36 (20 °C)
Flash Pt: 90 °C
CAS Number: 85272-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75e2d2906d4fa64713b4db3e45f74f7f5f0fa462&vid=75e2d2906d4fa64713b4db3e45f74f7f5f0fa462&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR