trans-N1,N2-Dimethylcyclohexane-1,2-diamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: trans-N,N'-Dimethyl-1,2-cyclohexanediamine

D2946-1G D2946-5G
TCD2946-1GEA 31.52 USD
TCD2946-1G TCD2946-5G
trans-N1,N2-Dimethylcyclohexane-1,2-diamine ≥98.0% (by titrimetric analysis)
trans-N1,N2-Dimethylcyclohexane-1,2-diamine

CAS Number: 67579-81-1
MDL Number: MFCD03001702
Molecular Formula: C8H18N2
Molecular Weight: 142.25
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 83
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: trans-1,2-Bis(methylamino)cyclohexane

Formula: C₈H₁₈N₂
MW: 142.24 g/mol
Boiling Pt: 83 °C
Density: 0.91 (20 °C)
MDL Number: MFCD03001702
CAS Number: 67579-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f74281b068649691a18389b4909426325747d3ca&vid=f74281b068649691a18389b4909426325747d3ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR