trans-1,4-Dibromo-2-butene ≥98.0%

Supplier: TCI America

Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene

D2944-250G D2944-25G
TCD2944-250GEA 645 USD
TCD2944-250G TCD2944-25G
trans-1,4-Dibromo-2-butene ≥98.0%
trans-1,4-Dibromo-2-butene

CAS Number: 821-06-7
MDL Number: MFCD00000249
Molecular Formula: C4H6Br2
Molecular Weight: 213.90
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 76
Melting point (°C): 54
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₄H₆Br₂
MW: 213.9
Boiling Pt: 203
Melting Pt: 54 °C
Flash Pt: 110 °C
MDL Number: MFCD00000249
CAS Number: 821-06-7
UN: 2923
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4e76fdf29a82a8a97148a15d94ffca2d0db937d&vid=d1e042df6760fc801521d29184bb71e623d2b9f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR