Dicyclohexyl(trifluoromethanesulfonyloxy)borane ≥97.0%

Supplier: TCI America

Synonyms: Trifluoromethanesulphonic acid Dicyclohexylboryl Ester, Dicyclohexylboron Triflate

D2943-1G D2943-5G
TCD2943-1GEA 69 USD
TCD2943-1G TCD2943-5G
Dicyclohexyl(trifluoromethanesulfonyloxy)borane ≥97.0%
Dicyclohexyl(trifluoromethanesulfonyloxy)borane

CAS Number: 145412-54-0
MDL Number: MFCD06797094
Molecular Formula: C13H22BF3O3S
Molecular Weight: 326.18
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Dicyclohexylboron Triflate, Trifluoromethanesulfonic Acid Dicyclohexylboryl Ester

Formula: C₁₃H₂₂BF₃O₃S
MW: 326.18 g/mol
MDL Number: MFCD06797094
CAS Number: 145412-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44b509c4f69670b7fac56a393381db9577385c96&vid=44b509c4f69670b7fac56a393381db9577385c96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR