3,4-Difluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,4-Difluorobenzene-1-sulphonyl chloride

D2941-1G D2941-5G
TCD2941-1GEA 42 USD
TCD2941-1G TCD2941-5G
3,4-Difluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3,4-Difluorobenzenesulphonyl chloride

CAS Number: 145758-05-0
MDL Number: MFCD00153120
Molecular Formula: C6H3ClF2O2S
Molecular Weight: 212.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.56

Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Density: 1.56 (20 °C)
MDL Number: MFCD00153120
CAS Number: 145758-05-0
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00d16e46bcb4a8b728f95f01ecf191ce2ed86d50&vid=00d16e46bcb4a8b728f95f01ecf191ce2ed86d50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR