2,4-Difluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,4-Difluorobenzene-1-sulphonyl chloride

D2939-25G D2939-5G
TCD2939-25GEA 204 USD
TCD2939-25G TCD2939-5G
2,4-Difluorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
2,4-Difluorobenzenesulphonyl chloride

CAS Number: 13918-92-8
MDL Number: MFCD00042481
Molecular Formula: C6H3ClF2O2S
Molecular Weight: 212.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 84
Specific Gravity (20/20): 1.57

Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 84 °C
Density: 1.57 (20 °C)
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4bf76fe2d8dbfdf36365597ec5834ecda1e9606&vid=f4bf76fe2d8dbfdf36365597ec5834ecda1e9606&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR