4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride, DMTMM, 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium chloride
D2919-25G
D2919-5G
TCD2919-25GEA
385.96
USD
TCD2919-25G
TCD2919-5G
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride ≥98.0% (by total nitrogen basis)
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride
CAS Number: 3945-69-5
MDL Number: MFCD03613550
Molecular Formula: C10H17ClN4O3
Molecular Weight: 276.72
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: DMTMM
Formula:
C₁₀H₁₇ClN₄O₃ MW: 276.72 g/mol |
MDL Number:
MFCD03613550 CAS Number: 3945-69-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...