2,6-Dichloro-4-nitroanisole ≥98.0% (by GC)

Supplier: TCI America

D2872-1G D2872-5G
TCD2872-1GEA 55.4 USD
TCD2872-1G TCD2872-5G
2,6-Dichloro-4-nitroanisole ≥98.0% (by GC)
2,6-Dichloro-4-nitroanisole

CAS Number: 17742-69-7
MDL Number: MFCD00061129
Molecular Formula: C7H5Cl2NO3
Molecular Weight: 222.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: 1,3-Dichloro-2-methoxy-5-nitrobenzene

Formula: C₇H₅Cl₂NO₃
Melting Pt: 100 °C
MDL Number: MFCD00061129
CAS Number: 17742-69-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0475ba3a14a0f12662660bcfe6cfbe2926bec830&vid=0475ba3a14a0f12662660bcfe6cfbe2926bec830&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR