3,4-Difluoroanisole ≥97.0% (by GC)

Supplier: TCI America

D2856-25G D2856-5G
TCD2856-25GEA 157 USD
TCD2856-25G TCD2856-5G
3,4-Difluoroanisole ≥97.0% (by GC)
3,4-Difluoroanisole

CAS Number: 115144-40-6
MDL Number: MFCD00010730
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 55
Specific Gravity (20/20): 1.24

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Density: 1.24 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00010730
CAS Number: 115144-40-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=893610bfb8a4f3be8f448d647253e55a9610e1cc&vid=5d3e1c18e3356a3c95f8c09c16acc9e5f31b1eab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR